Thermo Scientific STAR A326 Star A326 pH/DO Portable Meter Kit

Representative image only.

Thermo Scientific STAR A326 Star A326 pH/DO Portable Meter Kit

Thermo Scientific | Mfr: STARA3265| Item: 5882574

Advanced features and dependability makes it the top choice for measuring samples

$4,904.18CAD / Each

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$4,904.18CAD / Each

Buy It Together

Current Item

Thermo Scientific STAR A326 Star A326 pH/DO Portable Meter Kit
Thermo Scientific STAR A326 Star A326 pH/DO Portable Meter Kit
5882574

$4,904.18CAD / Each
+ =
Total Price 5177.86 ADD 2 ITEMS TO CART
Product Description

Includes

4 AA batteries.

Key Features

  • Multilanguage interface supports Engliah, Spanish, French, Italian, German and Chinese
  • Improved stability algorithm provide better measurement accuracy
  • Ensure temperature accuracy with ATC temperature calibration
  • Nonvolatile memory holds up to 5000 data points with time date stamp
  • Individual method passwords provide better security
  • Four AA batteries provide over 800 hours of field use, or order optional AC adapter for lab use
  • Choose between RDO optical probe and polarographic probe
  • Directly measure in mg/L (ppm) or % saturation
  • Automatic temperature, barometric pressure and salinity correction
  • 3 year warranty

More About this Item

Thermo Scientific Orion Star A 323 RDO/DO and A326 pH/RDO/DO meters features an intuitive user interface with plain language screen prompts. A large, backlit graphic LCD, shows measurement and temperature along with electrode status, time/date, sample ID, user ID, and calibration data.

GLP-related features include 5000 data points with time/date stamp and electrode verification. Meter utilizes RS-232 or USB for connectivity and includes data analysis software.

Available as meter only and as complete meter kit configurations

Kit includes ROSS™ pH/ATC electrode, RDO probe, 10 pouches each of 4, 7, 10 buffer and rinse solution, 475 mL of ROSS electrode storage solution, protective armor with electrode holder, hard carrying case, and four AA batteries.

3 Year WarrantyIP 67
Specifications
  • Meter TypePortable
  • Display typeGraphic LCD
  • ConnectionspH: BNC; Temperature: 8-pin mini-DIN
  • Min pH (pH)-2
  • Max pH (pH)20
  • pH Resolution0.001/0.01/0.1
  • pH Accuracy±0.002 pH
  • Buffer RecognitionDIN, NIST, and USA
  • CalibrationUp to 5 points
  • Min mV (mV)-2000
  • Max mV (mV)2000
  • mV Resolution0.1 or 1 mV
  • mV Accuracy±0.2 mV or 0.05%, whichever is greater
  • Min Dissolved Oxygen0 ppm; 0%
  • Max Dissolved Oxygen90 ppm; 600%
  • Min Temperature (° C)-5
  • Max Temperature (° C)105
  • Min Temperature (° F)23
  • Max Temperature (° F)221
  • InterfaceRS-232 and USB
  • WaterproofYes
  • Battery4 AA (included)
  • Battery life (hr)800
  • Width (cm)10.795
  • Height (cm)23.1775
  • Width (in)4 1/4
  • Length (in)2 3/8
  • Height (in)9 1/8
  • Probe IncludedYes
  • DescriptionStar A326 pH/DO Portable Meter Kit
  • Warranty3 year
Reviews
Q&A