Thermo Scientific STAR A216 Star A216 pH/DO Benchtop Meter Kit

Representative image only.

Thermo Scientific STAR A216 Star A216 pH/DO Benchtop Meter Kit

Thermo Scientific | Mfr: STARA2165| Item: 5882542

Durability and dependability make it the ideal choice for every lab, every sample, everyday

$6,276.42CAD / Each

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$6,276.42CAD / Each

Buy It Together

Current Item

Thermo Scientific STAR A216 Star A216 pH/DO Benchtop Meter Kit
Thermo Scientific STAR A216 Star A216 pH/DO Benchtop Meter Kit
5882542

$6,276.42CAD / Each
+ =
Total Price 6550.10 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Multilanguage interface supports English, Spanish, French, Italian, German and Chinese
  • Improved stability algorithm provide better measurement accuracy
  • Ensure temperature accuracy with ATC temperature calibration
  • Nonvolatile memory holds up to 2000 data points with time date stamp
  • Individual method passwords provide better security
  • Option of AC power or four AA batteries (sold separtely)
  • 3 year warranty

More About this Item

Thermo Scientific Orion Star A216 multi-parameter meters offer pH, conductibity and temperature as well as a pH, conductivity, dissolved oxygen and temperature meter. Meters features an intuitive user interface with plain language prompts. A large, backlit graphic LCD shows measurements with temperature along with electrode status, time/date, sample ID, user ID, and calibration data.
GLP-related features include 2000 data points with time/date stamp and electrode verification. Meter utilizes RS-232 or USB for connectivity and includes data analysis software.

Up to five point automatic calibration with USA/NIST and Din buffers, auto-buffer recognition, and recalibration feature to fix calibration errors.

Choose between RDO optivcal probe or a polarographic probe for oxygen measurements. Directly measure in mg/L (ppm) or % saturation with automatic temperature, barometric pressure and salinity corrections.

Available as meter only and as complete meter kit configurations.

Meter includes ROSS™ Triode pH/ATC electrode(05718-28), polarographic probe (53101-60), calibration sleeve, maintenance kit, 475-mL each of 4, 7, 10 buffer and electrode storage solution, swing-arm electrode holder and power adapter.

3 Year WarrantyIP54 RatedISO9001:2000 Certified Supplier
Specifications
  • Meter TypeBenchtop
  • Display typeGraphic LCD
  • Min pH (pH)-2
  • Max pH (pH)20
  • pH Resolution0.001/0.01/0.1
  • pH Accuracy±0.002 pH
  • Buffer RecognitionDIN, NIST, and USA
  • CalibrationUp to 5 points
  • Min mV (mV)-2000
  • Max mV (mV)2000
  • mV Resolution0.1 or 1 mV
  • mV Accuracy±0.2 mV or 0.05%, whichever is greater
  • Min Dissolved Oxygen0 to 90 ppm; 0 to 600 %
  • Dissolved Oxygen Accuracy±0.5% of reading
  • Temperature compensationAutomatic or manual
  • WaterproofYes
  • Width (cm)17.78
  • Height (cm)24.765
  • Width (in)7
  • Length (in)3 3/8
  • Height (in)9 3/4
  • DescriptionStar A216 pH/DO Benchtop Meter Kit
  • Warranty3 year
Reviews
Q&A