Thermo Scientific STAR A121 Star A121 pH Portable Meter Kit

Representative image only.

Thermo Scientific STAR A121 Star A121 pH Portable Meter Kit

Thermo Scientific | Mfr: STARA1215| Item: 5882508

Combine simplicity with accuracy for your portable pH measurements

$2,405.68CAD / Each

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$2,405.68CAD / Each

Buy It Together

Current Item

Thermo Scientific STAR A121 Star A121 pH Portable Meter Kit
Thermo Scientific STAR A121 Star A121 pH Portable Meter Kit
5882508

$2,405.68CAD / Each
+ + =
Total Price 3349.24 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Includes

4 AA batteries.

Key Features

  • Large LCD displays pH or mV along with temperature
  • Up to 3 point calibration, with easy recall of slope data
  • Auto-Read™ function locks in stable reading
  • Automatic recognition of USA/NIST and DIN buffers
  • Nonvolatile memory holds up to 50 data points
  • Waterproof, IP67-rated housing
  • Four AA batteries provide over 2000 hours of field use, or order optional AC adapter for lab use
  • 3 year warranty

More About this Item

Thermo Scientific Orion Star A121 portable pH meter is ideal for field testing--it features waterproof and durable IP67-rated housing. Large informative display shows pH and mV readings with temperature, battery life, electrode status, meter mode, calibration data, power source, and more. Simple button layout and on-screen messages help with calibration and setup menu choices.

Accessories include:Orion Triode™ pH/ATC electrode 58823-30; 60-mL of pH buffers 4, 7, 10 and storage solution; protective armor; hard carrying case; and four AA batteries.

3 Year WarrantyIP 67
Specifications
  • Meter TypePortable
  • Display typeLCD
  • Min pH (pH)-2
  • Max pH (pH)16
  • pH Resolution0.1/0.01
  • pH Accuracy±0.01 pH
  • Buffer RecognitionDIN, NIST, and USA
  • CalibrationUp to 3 points
  • Min mV (mV)-1999
  • Max mV (mV)1999
  • mV Resolution0.1/1
  • mV Accuracy±0.2 mV or 0.05%, whichever is greater
  • Dissolved Oxygen Accuracy±2% full scale
  • Min Temperature (° C)-5
  • Max Temperature (° C)105
  • Min Temperature (° F)23
  • Max Temperature (° F)221
  • Temperature Resolution0.1
  • Temperature Accuracy±0.1°C
  • Temperature compensationAutomatic or manual
  • InterfaceUSB or RS232
  • WaterproofYes
  • Battery4 AA (included)
  • Width (cm)10.795
  • Height (cm)23.1775
  • Width (in)4 1/4
  • Length (in)2 3/8
  • Height (in)9 1/8
  • DescriptionStar A121 pH Portable Meter Kit
  • Warranty3 year
Reviews
Q&A