Thermo Scientific Orion Star T910 pH Titrator without Electrode; 100 to 240 VAC

Representative image only.

Thermo Scientific Orion Star T910 pH Titrator without Electrode; 100 to 240 VAC

Thermo Scientific | Mfr: START9100| Item: 9410200

Get fast, reliable, and reproducible automated titrations

$5,908.29CAD / Each

$5,908.29CAD / Each

Buy It Together

Current Item

Thermo Scientific Orion Star T910 pH Titrator without Electrode; 100 to 240 VAC
Thermo Scientific Orion Star T910 pH Titrator without Electrode; 100 to 240 VAC
9410200

$5,908.29CAD / Each
+ =
Total Price 5998.02 ADD 2 ITEMS TO CART
Product Description

Includes

Stirrer probe, dispenser probe, 20-mL burette, standard tubing kit, 1-L plastic bottle, GL38 bottle cap with desiccant tube, computer cable, literature on USB drive, and 110 to 240 VAC power supply.

Key Features

  • Provides dedicated acid-base titrations
  • Program and save up to 10 user-defined methods
  • Store up to 100 titration data sets with date/time stamp
  • Easily transfer data to printer, computer, or USB drive
  • Minimize downtime with replaceable burettes, tubing, and dispensers
  • Large easy-to-ready touchscreen display enables fast setup
  • 1 year warranty

More About this Item

The T900 series automated potentiometric titrators feature a state-of-the-art reagent dispensing system designed to simplify your titrations and offer more reliable, reproducible, and faster results then manual titrations. These compact, rugged titrators are simple to use, let you easily navigate live titration analysis, and are ideal for small bench spaces. You can safely run the titrator unattended until titration is completed.

The automated titrant delivery system, featuring a high-accuracy burette and dispenser, provides consistent results every time. Internal data storage maintains titration history, titrant standardizations, electrode calibrations, and direct measurements, all of which can easily be transferred to an external device.

T910 pH Titrator is specially designed for acid-based titrations including titratable acidity of juices and wines, acidity of food products, alkalinity of water, acidity and alkalinity of consumer products, total acid number (TAN), and total base number (TBN). Titration techniques include equivalence point titrations and preset pH or mV endpoint titrations for versatile sample analysis.

1 Year WarrantyConformite Europeenne
Specifications
  • UnitspH, mV
  • Display TypeTouchscreen
  • Min pH (pH)-2
  • Max pH (pH)20
  • pH Resolution0.001, 0.01, 0.1
  • pH Accuracy±0.002
  • Min mV (mV)-2000
  • Max mV (mV)2000
  • mV Resolution0.1
  • mV Accuracy±0.2
  • Min Temperature (° C)-5
  • Max Temperature (DegreeC)100
  • Temperature Resolution0.1°F (0.1°C)
  • Temperature Accuracy±0.2°C
  • Capacity (mL)20
  • OutputUSB
  • Data LoggingYes
  • Memory Size100 data sets
  • Power (VAC)110 to 240
  • Power (Hz)50/60
  • Length (in)10
  • Width (in)16
  • Height (in)14
  • Length (cm)25.4
  • Width (cm)40.6
  • Height (cm)35.6
  • DescriptionpH Titrator without Electrode; 100 to 240 V
  • Warranty1 Year
Reviews
Q&A