Thermo Scientific Nunc 269620 Collection Plate for Silent Screen Plates

Representative image only.

Thermo Scientific Nunc 269620 Collection Plate for Silent Screen Plates

Thermo Scientific | Mfr: 269620| Item: 0192802

Surfaces with different degrees of hydrophilicity—ideal for solid phase immunoassays (ELISA)

$325.62CAD / Case of 60

$325.62CAD / Case of 60

Buy It Together

Current Item

Thermo Scientific Nunc 269620 Collection Plate for Silent Screen Plates
Thermo Scientific Nunc 269620 Collection Plate for Silent Screen Plates
0192802

$325.62CAD / Case of 60
+ =
Total Price 925.29 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Engineered for easy/reproducible passive absorption
  • Certified homogeneity of binding
  • Clear polystyrene has excellent optical properties making it ideal for optical measurements
  • Plates are optimized for robotics, plate readers and liquid handlers

More About this Item

Surface binding assay microplate wells facilitate surface adsorption of proteins, antibodies, and other biomolecules. The surface adsorption aids in applications for the basis for quantitative assays such as ELISA.

Choose from treated or untreated plates. Untreated polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment. Untreated surfaces are ideal for adding coatings in your lab. MaxiSorp surface treatment is hydrophilic and optimized for binding immunoglobulin but also binds a wide range of other proteins.

Specifications
  • Number of Wells96
  • Well Volume (mL)0.4
  • Min Working Well Volume (mL)0.05
  • Max Working Well Volume (mL)0.25
  • Well Bottom Diameter (mm)6.2
  • Well Top Diameter (mm)7
  • Well ShapeFlat
  • Surface TreatmentNon-Treated
  • Lid (Included)No
  • SterileNo
  • Plate ColorClear
  • MaterialPolystyrene
  • Height (mm)14.4
  • Width (mm)86
  • Length (mm)128
  • DescriptionSurface Binding Assay Microplates, 96-Well, Polystyrene, Non-Treated, Flat, 0.4 mL; 60/Cs
Reviews
Q&A