Thermo Scientific Nalgene DS0390-0070 Filling Bell for Liquid Transfer, Polycarbonate
Representative image only.
Thermo Scientific Nalgene DS0390-0070 Filling Bell for Liquid Transfer, Polycarbonate
Thermo Scientific | Mfr: DS0390-0070| Item: 0623920
Aseptic transfer of liquids and cell culture media
$98.42CAD / Each
BUY MORE & SAVE
  • 1-5$98.42 CAD / Each
  • 6-29$88.58 CAD / Each
  • 30-119$83.66 CAD / Each
  • 120+$78.74 CAD / Each
$98.42CAD / Each
BUY MORE & SAVE
  • 1-5$98.42 CAD / Each
  • 6-29$88.58 CAD / Each
  • 30-119$83.66 CAD / Each
  • 120+$78.74 CAD / Each
Product Description
Key Features
  • Aseptic transfer of liquids and media
  • Ideal for microbiology and cell culture applications
  • Measures 11.4 cm H x 7 cm ID (4-1/2" x 2-3/4") with adapter
  • 1 year warranty
More About this Item
This polycarbonate filling bell allows aseptic transfer of liquids and cell culture media— ideal for microbiology and cell culture applications. It is nontoxic, durable, and costs less than glass bells. Polypropylene adapter for 1/4" to 1/2" ID tubing is included. Filling tube measures 2-1/2"L x 3/8"OD (6.4 x 1.0 cm). Autoclavable.
1 Year Warranty
Specifications
  • Height (InchShort)4 31/64
  • ColorClear
  • MaterialPolycarbonate
  • SterileNo
  • Inside Diameter (ID) (mm)70
  • DescriptionFilling bell for liquid transfer, 4-1/2"H x 2-7/8"ID, PC