Thermo Scientific Nalgene 6432-0010 quick-action Tefzel ETFE spigot
Representative image only.
Thermo Scientific Nalgene 6432-0010 quick-action Tefzel ETFE spigot
Thermo Scientific | Mfr: 6432-0010| Item: 0607023
$297.62CAD / Each
BUY MORE & SAVE
  • 1-5$297.62 CAD / Each
  • 6-29$267.86 CAD / Each
  • 30-119$252.98 CAD / Each
  • 120+$238.10 CAD / Each
$297.62CAD / Each
BUY MORE & SAVE
  • 1-5$297.62 CAD / Each
  • 6-29$267.86 CAD / Each
  • 30-119$252.98 CAD / Each
  • 120+$238.10 CAD / Each
Buy It Together
Current Item
Thermo Scientific Nalgene 6432-0010 quick-action Tefzel ETFE spigot
Thermo Scientific Nalgene 6432-0010 quick-action Tefzel ETFE spigot
0607023
$297.62CAD / Each
+ + =
Total Price 1296.21 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 1 year warranty
More About this Item
Use these spigots with NALGENE carboys. Spigots have 1-1/8"-12 UNF threads and accept 5/8" tubing. Spigot 06073-22 is made of polypropylene and includes an adapter for 1/4" or 1/2" ID tubing. Spigot 06073-23 is made of TEFZEL ETFE; does not include a tubing adapter. NALGENE.
1 Year Warranty
Specifications
  • MaterialEthylene Tetrafluoroethylene (ETFE)
  • Cap MaterialEthylene Tetrafluoroethylene (ETFE)
  • AutoclavableYes
  • Descriptionquick-action Tefzel ETFE spigot