Thermo Scientific Nalgene 6420-0750 needle-type tank spigot, 3/4" NPT(M)
Representative image only.
Thermo Scientific Nalgene 6420-0750 needle-type tank spigot, 3/4" NPT(M)
| Mfr: 6420-0750| Item: 0607030
Instant on/off control ensures fast, smooth flow
$235.74CAD / Each
BUY MORE & SAVE
  • 1-5$235.74 CAD / Each
  • 6-29$212.17 CAD / Each
  • 30-119$200.38 CAD / Each
  • 120+$188.59 CAD / Each
$235.74CAD / Each
BUY MORE & SAVE
  • 1-5$235.74 CAD / Each
  • 6-29$212.17 CAD / Each
  • 30-119$200.38 CAD / Each
  • 120+$188.59 CAD / Each
Product Description
Key Features
  • Stem is LDPE
  • 1 year warranty
More About this Item
These faucets are manufactured of HDPE for greater strength and positive thread engagement.
1 Year Warranty
Specifications
  • MaterialLow Density Polyethyene (LDPE)
  • Cap MaterialLow-Density Polyethylene (LDPE)
  • Cap / Stopper Size3/4" NPT
  • Descriptionneedle-type tank spigot, 3/4" NPT(M)