Thermo Scientific Nalgene 6379-0010 Fluorinated HDPE Safety Waste System

Representative image only.

Thermo Scientific Nalgene 6379-0010 Fluorinated HDPE Safety Waste System

Thermo Scientific | Mfr: 6379-0010| Item: 8140014

Dispose of hazardous chemicals quickly and easily

$456.75CAD / Each

$456.75CAD / Each
Product Description

Key Features

  • 1 year warranty

More About this Item

System includes a waste bottle and funnel that screws onto bottle opening for quick disposal. The hinged lid on the funnel is sealed to protect against dangerous vapors; long funnel stem helps minimize evaporation.

Model 81400-14 is made of fluorinated HDPE for even greater chemical resistance.

ISO9001 Certified Supplier1 Year Warranty
Specifications
  • MaterialHigh Density Polyethyene (HDPE)
  • Capacity (Gal)2.64
  • Capacity (Liters)10
  • Funnel Mouth ID (in)10
  • Funnel Stem Length (in)10
  • Funnel Mouth ID (cm)25.4
  • Funnel Stem Length (cm)25.4
  • DescriptionFluorinated HDPE Safety Waste System
  • Warranty1 Year
Reviews
Q&A