Thermo Scientific Nalgene 3664-0250 Polypropylene Graduated Cylinder, 250 mL

Representative image only.

Thermo Scientific Nalgene 3664-0250 Polypropylene Graduated Cylinder, 250 mL

Thermo Scientific | Mfr: 3664-0250| Item: 0613552

$27.15CAD / Each
$38.31CAD / Each

$27.15CAD / Each
$38.31CAD / Each
Product Description

Key Features

  • Polypropylene (PP) cylinders offer good chemical resistance
  • Raised graduations allow easy reading and precise measuring
  • Large, ribbed base holds cylinder stable
  • Autoclavable
  • 1 year warranty
1 Year Warranty
Specifications
  • MaterialPolypropylene
  • Capacity (mL)250
  • Graduation Interval (mL)2
  • DescriptionPolypropylene Graduated Cylinder, 250 mL
  • Warranty1 year
Reviews
Q&A