Thermo Scientific Nalgene 2158-0032 Quick Filling/Venting Closures, PP, 83b 3-port 3/8"
Representative image only.
Thermo Scientific Nalgene 2158-0032 Quick Filling/Venting Closures, PP, 83b 3-port 3/8"
Thermo Scientific | Mfr: 2158-0032| Item: 6250812
$471.48CAD / Each
BUY MORE & SAVE
  • 1-5$471.48 CAD / Each
  • 6-29$424.33 CAD / Each
  • 30-119$400.76 CAD / Each
  • 120+$377.18 CAD / Each
$471.48CAD / Each
BUY MORE & SAVE
  • 1-5$471.48 CAD / Each
  • 6-29$424.33 CAD / Each
  • 30-119$400.76 CAD / Each
  • 120+$377.18 CAD / Each
Product Description
More About this Item
Ideal for filling/dispensing into and out of carboys and bottles with an 83B neck finish. Quick disconnect tubing ports make installation a snap. Order replacement straight hose barb inserts and optional elbow hose barb inserts below. Note: Polyproplene is autoclavable.
Specifications
  • MaterialPolypropylene(PP)
  • Cap MaterialPolypropylene (PP)
  • Cap / Stopper Size83B
  • Cap ColorWhite
  • Tubing ID9.5 mm
  • DescriptionQuick Filling/Venting Closures, PP, 83b 3-port 3/8"