Thermo Scientific Nalgene 2158-0031 Quick Filling/Venting Closures, PP, 83b 3-port 1/4"
Representative image only.
Thermo Scientific Nalgene 2158-0031 Quick Filling/Venting Closures, PP, 83b 3-port 1/4"
Thermo Scientific | Mfr: 2158-0031| Item: 6250810
$483.27CAD / Each
BUY MORE & SAVE
  • 1-5$483.27 CAD / Each
  • 6-29$434.94 CAD / Each
  • 30-119$410.78 CAD / Each
  • 120+$386.62 CAD / Each
$483.27CAD / Each
BUY MORE & SAVE
  • 1-5$483.27 CAD / Each
  • 6-29$434.94 CAD / Each
  • 30-119$410.78 CAD / Each
  • 120+$386.62 CAD / Each
Product Description
More About this Item
Ideal for filling/dispensing into and out of carboys and bottles with an 83B neck finish. Quick disconnect tubing ports make installation a snap. Order replacement straight hose barb inserts and optional elbow hose barb inserts below. Note: Polyproplene is autoclavable.
Specifications
  • MaterialPolypropylene(PP)
  • Cap MaterialPolypropylene (PP)
  • Cap / Stopper Size83B
  • Cap ColorWhite
  • Tubing ID6.4 mm
  • DescriptionQuick Filling/Venting Closures, PP, 83b 3-port 1/4"