Thermo Scientific Nalgene 127-0020 Filter Units, Gridded 0.2um CN, 1000 mL; 12/Pk

Representative image only.

Thermo Scientific Nalgene 127-0020 Filter Units, Gridded 0.2um CN, 1000 mL; 12/Pk

Thermo Scientific | Mfr: 127-0020| Item: 0673092

Provide fast flow rates for buffers, salts, and other aqueous solutions

$677.98CAD / Pkg of 12

$677.98CAD / Pkg of 12
Product Description

Key Features

  • Triton-free CN membranes have excellent wetting properties — ideal for filtering and clarifying solutions when protein binding is not a concern

More About this Item

Nalgene® Filter Units provide a complete vacuum filtration system designed for the filtration of cell culture media, biological fluids, and other aqueous solutions. Each filter unit consists of polystyrene housing with a welded filter membrane mounted to a support plate in the upper structure. Slide arms with quick-disconnect tubing adapters simplify vacuum line attachment. The upper structures are easily separated from their receivers and a sterile, leakproof closure is included for filtrate storage.

Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.

Sterile
Specifications
  • Membrane MaterialCellulose Nitrate
  • Pore Size (µm)0.2
  • Membrane Diameter (mm)90
  • Housing MaterialPolystyrene (PS)
  • Capacity (mL)1000
  • SterileYes
  • DescriptionFilter Units, Gridded 0.2um CN, 1000 mL; 12/Pk
Reviews
Q&A