Thermo Scientific GENESYS™ Test Tube Holder for GENESYS™ 30, 40, and 50
Representative image only.
Thermo Scientific GENESYS™ Test Tube Holder for GENESYS™ 30, 40, and 50
Thermo Scientific | Mfr: 840-277500| Item: 0265460
Enhance the versitility of your spectrophotometer
$519.74CAD / Each
$519.74CAD / Each
Buy It Together
Current Item
Thermo Scientific GENESYS™ Test Tube Holder for GENESYS™ 30, 40, and 50
Thermo Scientific GENESYS™ Test Tube Holder for GENESYS™ 30, 40, and 50
0265460
$519.74CAD / Each
+ =
Total Price 6545.91 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Expand your spectrophotometer capacity and capabilities
  • 2 year warranty
More About this Item
Choose from a wide range of replacement parts and accessories including cuvette holders, sipper pumps, and more suitable for use with the Thermo Scientific™ GENESYS™ visible and UV-visible spectrophotometers. Spectrophotometers sold separately.
This test tube holder accomidates tubes and vials with an outer diameter of 12 to 25 mm.
Available for sale and use in the United States only.
Two year warranty
Specifications
  • Compatible WithGENESYS 30, 40, and 50 spectrophotometers
  • DescriptionTest Tube Holder for GENESYS™ 30, 40, and 50