Thermo Scientific GENESYS™ Long Path 5 to 100 mm Cylindrical Cell Holder for GENESYS™ 30, 40, and 50
Representative image only.
Thermo Scientific GENESYS™ Long Path 5 to 100 mm Cylindrical Cell Holder for GENESYS™ 30, 40, and 50
Thermo Scientific | Mfr: 840-277800| Item: 0265463
Enhance the versitility of your spectrophotometer
$365.77CAD / Each
$365.77CAD / Each
Buy It Together
Current Item
Thermo Scientific GENESYS™ Long Path 5 to 100 mm Cylindrical Cell Holder for GENESYS™ 30, 40, and 50
Thermo Scientific GENESYS™ Long Path 5 to 100 mm Cylindrical Cell Holder for GENESYS™ 30, 40, and 50
0265463
$365.77CAD / Each
+ =
Total Price 6391.94 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Expand your spectrophotometer capacity and capabilities
  • 2 year warranty
More About this Item
Choose from a wide range of replacement parts and accessories including cuvette holders, sipper pumps, and more suitable for use with the Thermo Scientific™ GENESYS™ visible and UV-visible spectrophotometers. Spectrophotometers sold separately.
Long path cylindrical cell holder is suitable for 5 to 100 mm pathlength cuvettes.
Available for sale and use in the United States only.
Two year warranty
Specifications
  • Compatible WithGENESYS 30, 40, and 50 spectrophotometers
  • DescriptionLong Path 5 to 100 mm Cylindrical Cell Holder for GENESYS™ 30, 40, and 50