Thermo Scientific GENESYS™ Film Filter Holder for GENESYS™ 30, 40, and 50
Representative image only.
Thermo Scientific GENESYS™ Film Filter Holder for GENESYS™ 30, 40, and 50
Thermo Scientific | Mfr: 840-278100| Item: 0265464
Enhance the versitility of your spectrophotometer
$614.77CAD / Each
$614.77CAD / Each
Buy It Together
Current Item
Thermo Scientific GENESYS™ Film Filter Holder for GENESYS™ 30, 40, and 50
Thermo Scientific GENESYS™ Film Filter Holder for GENESYS™ 30, 40, and 50
0265464
$614.77CAD / Each
+ =
Total Price 6640.94 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Expand your spectrophotometer capacity and capabilities
  • 2 year warranty
More About this Item
Choose from a wide range of replacement parts and accessories including cuvette holders, sipper pumps, and more suitable for use with the Thermo Scientific™ GENESYS™ visible and UV-visible spectrophotometers. Spectrophotometers sold separately.
This holder accomideas filters and lenses up to 50 mm long by 80 mm tall by 10 mm thick.
Available for sale and use in the United States only.
Two year warranty
Specifications
  • Compatible WithGENESYS 30, 40, and 50 spectrophotometers
  • DescriptionFilm Filter Holder for GENESYS™ 30, 40, and 50