Thermo Scientific E3450 Pura -Lite Resistivity Indicators, 50 k-ohm, 120 VAC
Representative image only.
Thermo Scientific E3450 Pura -Lite Resistivity Indicators, 50 k-ohm, 120 VAC
Thermo Scientific | Mfr: E3450| Item: 0150436
A low-cost alternative to digital purity meters
$493.59CAD / Each
$493.59CAD / Each
Product Description
Key Features
  • Designed for use with B-Pure systems
  • 1 year warranty
More About this Item
These indicators can be mounted directly on top of single- or doublecartridge housings with 1⁄2" NPT(F) connections.
Green light indicates water resistivity is above the threshold; red light warns if resistivity falls below threshold. Operates on 120 VAC power.
1 Year Warranty
Specifications
  • Inlet Port1/2" NPT(F)
  • Resistivity Threshold50 kΩ
  • DescriptionPura -Lite Resistivity Indicators, 50 k-ohm, 120 VAC