Thermo Scientific CODS01 COD Standard, 1000 ppm, Nonhazardous, 475 mL.

Representative image only.

Thermo Scientific CODS01 COD Standard, 1000 ppm, Nonhazardous, 475 mL.

Thermo Scientific | Mfr: CODS01| Item: 9958180

Check operations prior to using the instruments

$154.56CAD / Each

$154.56CAD / Each
Product Description

Key Features

  • Use with AQUAfast Colorimeters such as AQ4000 and AQ2040
  • 1000 ppm concentration

More About this Item

Get the most out of your colorimeters using calibration standards.

Specifications
  • Capacity (mL)475
  • Compatible WithThermo Scientific Orion COD Reactor & Colorimeter
  • DescriptionCOD Calibration Standard, 1000 ppm, Nonhazardous, 475 mL.
Documents & Certificates
Reviews
Q&A