Thermo Scientific Cimarec 50094713 Maxi Direct, with power supply, 100-240 V

Representative image only.

Thermo Scientific Cimarec 50094713 Maxi Direct, with power supply, 100-240 V

Thermo Scientific | Mfr: 50094713| Item: 0465150

Power setting are pre-selectable to optimize stirring efficiency for different liquid volumes

$1,944.87CAD / Each

$1,944.87CAD / Each
Product Description

Includes

6-ft power cord.

Key Features

  • Generates a powerful magnetic field for smooth stirring of large volumes and highly viscous media
  • Automatic start-up for safe stirrer acceleration
  • Memory function stores last speed settings
  • 5 year warranty

More About this Item

Use this stirrer for general laboratory applications as well as for growing microorganisms, dissolving nutrient mediums and solids, and titration applications. Rugged, sturdy stirrer accommodates a variety of vessel shapes including Erlenmeyer flasks and beakers up to 5L. Durable, sealed stainless steel housing cleans easily under running water. Stirring speed remains constant even if viscosity of the medium changes. Stirrer is not for use in hazardous areas; not immersible.

5 Year Warranty
Specifications
  • Low Speed (rpm)80
  • High Speed (rpm)2000
  • No. Of Stirring Positions1
  • Max Stirring Volume (Liters)5
  • Length (in)8 29/64
  • Width (in)7 3/32
  • Height (in)1 3/8
  • Length (cm)21.49
  • Width (cm)18
  • Height (cm)3.5
  • Power (VAC)100 to 240
  • Power (Hz)50/60
  • DescriptionMaxi Direct, with power supply, 100-240 V
  • Warranty5 years
Reviews
Q&A