Thermo Scientific Biobasic 73105-152130 AX HPLC Column, Polyethyleneimine, 5 µm, 150 mm L x 2.1 mm ID

Representative image only.

Thermo Scientific Biobasic 73105-152130 AX HPLC Column, Polyethyleneimine, 5 µm, 150 mm L x 2.1 mm ID

Thermo Scientific | Mfr: 73105-152130| Item: 0190144

Stable polymeric coating is suitable for a wide range of analyte separations

$2,276.39CAD / Each

$2,276.39CAD / Each
Product Description

Key Features

  • Weak anion exchange for multiple charged analytes
  • Can be used for hydrophilic interaction chromatography (HILIC)
  • Proprietary, protein-friendly, ion exchange ligands of polyethyleneimine (PEI) bound to pure deactivated silica
  • Low hydrophobicity
  • 1 month warranty

More About this Item

Thermo Scientific™ BioBasic™ HPLC columns provide improved performance for biological samples including peptides, proteins, and other biomolecules. The high purity silica and tailored pore sizes provide superior performance in terms of improved resolution, efficiency, reproducibility, and column life.

BioBasic AX Columns utilize a polymeric secondary-tertiary amine (polyethyleneimine) ligand that is bound to a highly pure, base-deactivated silica that provides separation of proteins, peptides, other anionic species, and polar molecules. The polyethyleneimine bonded phase allows for additional chromatographic options, including hydrophilic interaction chromatography (HILIC) — ideal for separation of sugars and other highly polar species. BioBasic AX columns are i

1 Month Warranty
Specifications
  • PhasePolyethyleneimine (PEI)
  • Column Length (mm)150
  • Column Inside Diameter (ID) (mm)2.1
  • Column Particle Size (µm)5
  • Tubing Inside Diameter (ID) (mm)2.1
  • DescriptionAX HPLC Column, Polyethyleneimine, 5 µm, 150 mm L x 2.1 mm ID
  • Warranty1 month
Reviews
Q&A