Thermo Scientific Biobasic 72305-254630 4 HPLC Column, C4, 5 µm, 250 mm L x 4.6 mm ID

Representative image only.

Thermo Scientific Biobasic 72305-254630 4 HPLC Column, C4, 5 µm, 250 mm L x 4.6 mm ID

Thermo Scientific | Mfr: 72305-254630| Item: 0189897

Provide excellent analyses of larger proteins and peptides

$2,070.11CAD / Each

$2,070.11CAD / Each
Product Description

Key Features

  • Lower carbon loading for retention of larger proteins and peptides
  • Based on 300Å silica for outstanding biomolecule performance
  • Outstanding reproducibility, efficiency and column lifetime
  • Excellent for LC/MS separations
  • 1 month warranty

More About this Item

Thermo Scientific™ BioBasic™ HPLC columns provide improved performance for biological samples including peptides, proteins, and other biomolecules. The high purity silica and tailored pore sizes provide superior performance in terms of improved resolution, efficiency, reproducibility, and column life.

BioBasic 4 Columns contain 300Å pore size silica with lower carbon loading, ideally suited for high-hydrophobicity peptides nominally greater than 5,000 Da. Provide less retention of hydrophobic peptides due to the shorter alkyl length.

1 Month Warranty
Specifications
  • PhaseC4
  • Column Length (mm)250
  • Column Inside Diameter (ID) (mm)4.6
  • Column Particle Size (µm)5
  • Tubing Inside Diameter (ID) (mm)4.6
  • Description4 HPLC Column, C4, 5 µm, 250 mm L x 4.6 mm ID
  • Warranty1 month
Reviews
Q&A