Thermo Scientific Biobasic 72105-154630 18 HPLC Column, C18, 5 µm, 150 mm L x 4.6 mm ID

Representative image only.

Thermo Scientific Biobasic 72105-154630 18 HPLC Column, C18, 5 µm, 150 mm L x 4.6 mm ID

Thermo Scientific | Mfr: 72105-154630| Item: 0189759

Provide excellent separation of small to medium peptides

$2,467.93CAD / Each

$2,467.93CAD / Each
Product Description

Key Features

  • High purity silica and stable bonding chemistry — ideal for Life Science applications
  • 300Å pore size for maximum performance with biomolecules
  • High peak capacity stationary phase
  • Outstanding reproducibility, efficiency and column lifetime
  • Excellent for LC/MS separations
  • 1 month warranty

More About this Item

Thermo Scientific™ BioBasic™ HPLC columns provide improved performance for biological samples including peptides, proteins, and other biomolecules. The high purity silica and tailored pore sizes provide superior performance in terms of improved resolution, efficiency, reproducibility, and column life.

BioBasic 18 Columns provide reversed phase separation of small to medium peptides. Columns contain 300Å pore size silica, which is ideally suited for medium- to low-hydrophobicity peptides less than 5000 Da. BioBasic 18 HPLC columns provide a highly stable, reproducible surface with sharp peak shape due to the dense bonding chemistry and surface deactivation.

1 Month Warranty
Specifications
  • PhaseC18
  • Column Length (mm)150
  • Column Inside Diameter (ID) (mm)4.6
  • Column Particle Size (µm)5
  • Tubing Inside Diameter (ID) (mm)4.6
  • Description18 HPLC Column, C18, 5 µm, 150 mm L x 4.6 mm ID
Reviews
Q&A