Thermo Scientific Barnstead 50132374 MicroPure UV Reservoir Feed

Representative image only.

Thermo Scientific Barnstead 50132374 MicroPure UV Reservoir Feed

Thermo Scientific | Mfr: 50132374| Item: 9927131

18.2 MΩ-cm water in a compact system

$11,433.51CAD / Each

$11,433.51CAD / Each

Buy It Together

Current Item

Thermo Scientific Barnstead 50132374 MicroPure UV Reservoir Feed
Thermo Scientific Barnstead 50132374 MicroPure UV Reservoir Feed
9927131

$11,433.51CAD / Each
+ =
Total Price 13201.58 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Feed water monitoring for maximizing cartridge life
  • Variable flow control knob for dispensing
  • Quick-change cartridges for rapid cartridge change, even mid-operation
  • Optional integrated 6-L feed water reservoir for areas without direct access to a water line
  • 1 year warranty

More About this Item

Barnstead MicroPure systems deliver up to 15 L/day of Type I ultrapure water from a pretreated feed water supply. Water product quality exceeds international standards ASTM Type I, ISO 3696 Grade 1, ASTM D1193 and CLSI-CLRW.

These compact systems are an economical option for a wide range of applications needs from basic buffer preparations to highly sensitive protocols such as HPLC, PCR, or applications that require precise TOC removal and measurement.

The ergonomically designed controller is backlit and tilts for easy operation and better visibility. Dispenser allows drop by drop or quick dispensing.

Recirculation pump prevents bacterial growth during standstill and allows for constant monitoring of all system functions, allowing for the immediate display of fault messages if a problem is detected.


Applications: Molecular biology, microbology, PCR,
IVF, monoclonal antibodies

Systems with UF option include an ultrafilter to remove endotoxins.

NOTE: If feed water is pretreated using RO or DI, system requires DI cartridge 99299-00 (order separately).

1 Year WarrantyConformite Europeenne
Specifications
  • ASTM Water GradeType I
  • Max Flow Rate (L/min)2
  • Inlet Port3/4" ID hosebarb
  • With UV LampNo
  • With UltrafilterYes
  • Source6-L feed reservoir
  • Feed Water Requirements (Source)potable tap water; pretreated by reverse osmosis, ion exchange, or distillation
  • Type I Water (Resistivity)18.2 MΩ-cm
  • Feed Water Requirements (Conductivity)<2 µS/cm
  • Type I Water (TOC)5 to 10 ppb
  • Feed Water Requirements (TOC)50 ppb, max
  • Feed Water Requirements (turbidity)1.0 NTU
  • Type I Water (Conductivity)0.055 µS/cm
  • Feed Water Requirements (Bacteria)<100 CFU/mL
  • Type I Water (Bacteria)<1
  • Type I Water (Endotoxins)<0.001
  • Feed Water Requirements (pressure)1.4 to 87 psi (0.1 to 6 bar)
  • Feed Water Requirements (Temperature)2-35⁰C
  • DescriptionMicroPure UV Reservoir Feed
  • Warranty1 year
Reviews
Q&A