Thermo Scientific Barnstead 50132373 MicroPure water system, UV

Representative image only.

Thermo Scientific Barnstead 50132373 MicroPure water system, UV

Thermo Scientific | Mfr: 50132373| Item: 9927130

18.2 MΩ-cm water in a compact system

$10,917.82CAD / Each

$10,917.82CAD / Each

Buy It Together

Current Item

Thermo Scientific Barnstead 50132373 MicroPure water system, UV
Thermo Scientific Barnstead 50132373 MicroPure water system, UV
9927130

$10,917.82CAD / Each
+ =
Total Price 12685.89 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Feed water monitoring for maximizing cartridge life
  • Variable flow control knob for dispensing
  • Quick-change cartridges for rapid cartridge change, even mid-operation
  • Optional integrated 6-L feed water reservoir for areas without direct access to a water line
  • 1 year warranty

More About this Item

Barnstead MicroPure systems deliver up to 15 L/day of Type I ultrapure water from a pretreated feed water supply. Water product quality exceeds international standards ASTM Type I, ISO 3696 Grade 1, ASTM D1193 and CLSI-CLRW.

These compact systems are an economical option for a wide range of applications needs from basic buffer preparations to highly sensitive protocols such as HPLC, PCR, or applications that require precise TOC removal and measurement.

The ergonomically designed controller is backlit and tilts for easy operation and better visibility. Dispenser allows drop by drop or quick dispensing.

Recirculation pump prevents bacterial growth during standstill and allows for constant monitoring of all system functions, allowing for the immediate display of fault messages if a problem is detected.


Applications: Molecular biology, microbology, PCR,
IVF, monoclonal antibodies

Systems with UF option include an ultrafilter to remove endotoxins.

NOTE: If feed water is pretreated using RO or DI, system requires DI cartridge 99299-00 (order separately).

1 Year WarrantyConformite Europeenne
Specifications
  • Length (")12
  • ASTM Water GradeType I
  • Capacity90 L/hr
  • Width (InchShort)11 51/64
  • Height (InchShort)21 29/64
  • Max Production (L/hour)1.5
  • Max Flow Rate (L/min)1.5
  • Max Flow Rate (L/hour)90
  • Max Flow Rate (GPH)23.8
  • Inlet PortR 3/4"
  • With UV LampNo
  • With UltrafilterYes
  • Resistivity Threshold18.2 MΏ-cm
  • TOC Monitor1 to 5ppb
  • Conductivity Threshold0.055 µS/cm
  • SourcePretreated feed water
  • TOC Threshold5ppb
  • Feed Water Requirements (Source)potable tap water; pretreated by reverse osmosis, ion exchange, or distillation
  • Type I Water (Resistivity)18.2 MΩ-cm
  • Type I Water (TOC)1 to 5 ppb
  • Feed Water Requirements (TOC)1 to 5 ppb
  • Type I Water (Conductivity)0.055 µS/cm
  • Feed Water Requirements (Bacteria)<0.1 CFU/mL
  • Type I Water (Bacteria)<0.1 CFU/mL
  • Type I Water (Endotoxins)<0.001
  • Feed Water Requirements (pressure)1.45 to 87 psi (0.1 to 6 bar)
  • Feed Water Requirements (Temperature)2-35⁰C
  • Max Inlet Water Temperature (° F)95
  • Min Inlet Water Temperature (° F)35.6
  • Max Inlet Water Temperature (° C)35
  • Min Inlet Water Temperature (° C)2
  • Min Inlet Pressure (PSI)1.45
  • Max Inlet Pressure (PSI)87
  • Min Inlet Pressure (bar)0.1
  • Max Inlet Pressure (bar)6
  • Min Operating Pressure (PSI)1.45
  • Max Operating Pressure (PSI)87
  • Min Operating Pressure (bar)0.1
  • Max Operating Pressure (bar)6
  • Pressure (bar)6
  • Pressure (PSI)87
  • Height (in)21.46
  • Depth (in)11.8
  • Width (in)11.8
  • Height (cm)54.5
  • Depth (cm)30
  • Width (cm)30
  • DescriptionMicroPure water system, UV
  • Warranty1 year
Reviews
Q&A