Thermo Scientific Barnstead 50131217 GenPure water system, UF/UV

Representative image only.

Thermo Scientific Barnstead 50131217 GenPure water system, UF/UV

Thermo Scientific | Mfr: 50131217| Item: 9928730

Integrated feed water monitoring for maximum efficiency and consistent quality

$20,922.14CAD / Each

$20,922.14CAD / Each

Buy It Together

Current Item

Thermo Scientific Barnstead 50131217 GenPure water system, UF/UV
Thermo Scientific Barnstead 50131217 GenPure water system, UF/UV
9928730

$20,922.14CAD / Each
+ =
Total Price 22690.21 ADD 2 ITEMS TO CART
Product Description

Includes

ultrapure polisher cartridge, sterile 0.2 ìm filter, pressure regulator, wall bracket, and power cord.

Key Features

  • Push-button operation for easy dispensing
  • Ergonomic controller tilts for optimal key pressing and easy readability
  • Digital microprocessor control automatically monitors and stores faults from the last four weeks
  • Quick-change cartridge design simplifies self maintenance, even in mid-operation
  • 1 year warranty

More About this Item

Barnstead GenPure systems deliver up to 200 L/day of Type I ultrapure water from a pretreated feed water supply. Water product quality exceeds international standards ASTM Type I, ISO 3696 Grade 1, ASTM D1193 and CLSI-CLRW. These economical systems are fully customizable to meet a wide range of applications needs from basic buffer preparations to highly sensitive protocols such as HPLC, PCR, or
applications that require precise TOC removal and measurement.

Ergonomically designed controller tilts for easy operation and better visibility. Illuminated four-line alphanumeric display shows important system parameters including conductivity/resistivity measurements and TOC value. The flexible dispenser offers a radius of 24" (60 cm) from the system for filling larger vessels or glassware washing.

The RS-232 interface with adjustable send-interval safely transfers all measured data, faults, date, and time to a computer or log printer. Data printing at preprogrammed intervals satisfies GLP guidelines; order external printer separately. Real-time clock and code-protected operating system prevents unauthorized changes to system settings. USP-compliant conductivity measurement with temperature
compensation can be switched on or off.


Applications: Molecular biology, PCR, DNA, monoclonal antibodies, cell culture media
Systems with UV option offer UV oxidation (185/254 nm) for reducing the content of microorganisms and their metabolites. Systems with UF option include an ultrafilter to remove endotoxins.

NOTE: If feed water is pretreated using RO or DI, system
requires DI cartridge 99299-00 (order separately).

Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.

1 Year WarrantyConformite Europeenne
Specifications
  • ASTM Water GradeType I
  • Max Flow Rate (L/min)2
  • Inlet Port3/4" ID hosebarb
  • With UV LampYes
  • With UltrafilterYes
  • Feed Water Requirements (Source)potable tap water; pretreated by reverse osmosis, ion exchange, or distillation
  • Type I Water (Resistivity)18.2 MΩ-cm
  • Feed Water Requirements (Conductivity)<2 µS/cm
  • Type I Water (TOC)1 to 5 ppb
  • Feed Water Requirements (TOC)50 ppb, max
  • Feed Water Requirements (turbidity)1.0 NTU
  • Type I Water (Conductivity)0.055 µS/cm
  • Feed Water Requirements (Bacteria)<100 CFU/mL
  • Type I Water (Bacteria)<1 CFU/mL
  • Type I Water (Endotoxins)<0.001 EU/mL
  • Feed Water Requirements (pressure)1.4 to 87 psi (0.1 to 6 bar)
  • Feed Water Requirements (Temperature)2-35⁰C
  • DescriptionGenPure water system, UF/UV
  • Warranty1 year
Reviews
Q&A