Thermo Scientific AC4P71 Free Chlorine Reagent, 100 Pk
Representative image only.
Thermo Scientific AC4P71 Free Chlorine Reagent, 100 Pk
Thermo Scientific | Mfr: AC4P71| Item: 9957052
Powder chemistries for AQ3070 colorimeter
$61.74CAD / Pkg of 1
$61.74CAD / Pkg of 1
Product Description
Includes
100 packs
Key Features
  • USEPA approved DPD method
  • Individula foil packs of 100 chemistries
More About this Item
Thermo Scientific quality reagents are compatible with Orbeco, Hach, and Chemetrics chemistries.
EPA United States Environmental protection agency
Specifications
  • Test ParametersFree Chlorine
  • Number of Tests Included100
  • DescriptionFree Chlorine Reagent, 100 Pk