Thermo Scientific 88880005 RT2 Advanced Stirring Hot Plate, Ceramic Top; 230V

Representative image only.

Thermo Scientific 88880005 RT2 Advanced Stirring Hot Plate, Ceramic Top; 230V

Thermo Scientific | Mfr: 88880005| Item: 8657607

Accurate PT100 sensor provides real-time temperature detection for sensitive experimental procedures

$2,269.02CAD / Each

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$2,269.02CAD / Each

Buy It Together

Current Item

Thermo Scientific 88880005 RT2 Advanced Stirring Hot Plate, Ceramic Top; 230V
Thermo Scientific 88880005 RT2 Advanced Stirring Hot Plate, Ceramic Top; 230V
8657607

$2,269.02CAD / Each
+
Thermo Scientific 88880149 3 Prong Support Clamp
8657694

$39.49CAD / Each
+ =
Total Price 2871.34 ADD 3 ITEMS TO CART
Product Description

Includes

temperature probe (PT 100, SN-8-4 connector sensor) and power cord with plug (230 VAC model includes cord set with plugs for China, Europe, and UK).

Key Features

  • Ceramic-coated aluminum top provides excellent temperature uniformity and is corrosion resistant
  • Overheat prevention circuit turns off the heater if the top plate temperature reaches 842°F (450°C)
  • "Hot top" warning visually alerts lab personnel of potential hazard
  • Digital control with PID microprocessor for easy operation
  • Three user-selectable temperature control modes: optimal, fast, and slow
  • Two-user selective timer; showing actual and set value simultaneously
  • User-controlled temperature limit setting
  • Low-profile design saves valuable bench space
  • Optional nonslip heating bath and transparent safety shield (sold separately)
  • Optional supporting rod and clamps (sold separately) allow flexibility in configuring other test equipment and accessories
  • 2 year warranty
Two year warrantyConformite Europeenne
Specifications
  • Top Plate Diameter (in)5 1/2
  • Top Plate Diameter (cm)13.97
  • Top Plate MaterialCeramic-coated Aluminum
  • Min Temperature (° F)75
  • Max Temperature (° F)662
  • Min Temperature (DegreeC)20
  • Max Temperature (° C)350
  • Low Speed (rpm)30
  • High Speed (rpm)2000
  • No. Of Stirring Positions1
  • Max Stirring Volume (Liters)20
  • Length (in)11 2/5
  • Width (in)6 19/64
  • Height (in)3 29/32
  • Length (cm)29
  • Width (cm)16
  • Height (cm)10
  • Plug TypeEuropean, UK, China
  • Power (VAC)230
  • DescriptionRT2 Advanced Stirring Hot Plate, Ceramic Top; 230V
  • Warranty2 year
Reviews
Q&A