Thermo Scientific 88880002 RT2 Basic Digital Hot Plate, Ceramic Top; 120V

Representative image only.

Thermo Scientific 88880002 RT2 Basic Digital Hot Plate, Ceramic Top; 120V

Thermo Scientific | Mfr: 88880002| Item: 8657601

Safely and accurately heat your samples

$1,591.26CAD / Each

for an additional
$356.56CAD / Each
for an additional
$481.80CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$1,591.26CAD / Each

Buy It Together

Current Item

Thermo Scientific 88880002 RT2 Basic Digital Hot Plate, Ceramic Top; 120V
Thermo Scientific 88880002 RT2 Basic Digital Hot Plate, Ceramic Top; 120V
8657601

$1,591.26CAD / Each
+ + =
Total Price 2175.31 ADD 3 ITEMS TO CART
Product Description

Includes

power cord with plug (230 VAC model includes cord set with plugs for China, Europe, and UK).

Key Features

  • Sleek low-profile design saves valuable bench space
  • Ceramic-coated aluminum top provides excellent temperature uniformity and is corrosion resistant
  • Digital control with PID microprocessor
  • "Hot top" warning visually alerts lab personnel of potential hazard even when unit is shut off
  • Built-in overtemperature shuts off if top plate temperature reaches 842°F (450°C)
  • Three user-selectable temperature control modes: optimal, fast, and slow
  • Display allows you to simultaneously view set point and actual temperature
  • Optional nonslip heating bath and transparent safety shield (sold separately)
  • 2 year warranty
Two year warrantyConformite Europeenne
Specifications
  • Top Plate Diameter (in)5 1/2
  • Top Plate Diameter (cm)13.97
  • Top Plate MaterialCeramic-coated Aluminum
  • Min Temperature (° F)75
  • Max Temperature (° F)662
  • Min Temperature (DegreeC)20
  • Max Temperature (° C)350
  • Max Load Capacity (lbs)55
  • Max Load Capacity (kg)25
  • Length (in)11 2/5
  • Width (in)6 19/64
  • Height (in)3 29/32
  • Length (cm)28.956
  • Width (cm)16.002
  • Height (cm)9.906
  • Power (VAC)120
  • Power (Hz)60
  • Power (watts)600
  • DescriptionRT2 Basic Digital Hot Plate, Ceramic Top; 120V
  • Warranty2 year
Reviews
Q&A