Thermo Scientific 50129887PR Smart2Pure 6 l/h UV/UF, complete package

Representative image only.

Thermo Scientific 50129887PR Smart2Pure 6 l/h UV/UF, complete package

Thermo Scientific | Mfr: 50129887| Item: 9925914

Both Type I and Type II water in a complete space-saving design

$18,122.70CAD / Each

$18,122.70CAD / Each

Buy It Together

Current Item

Thermo Scientific 50129887PR Smart2Pure 6 l/h UV/UF, complete package
Thermo Scientific 50129887PR Smart2Pure 6 l/h UV/UF, complete package
9925914

$18,122.70CAD / Each
+ + + =
Total Price 19744.31 ADD 4 ITEMS TO CART
Product Description

Includes

RO/pretreatment cartridge, ultrapure polisher cartridge, sterile 0.2 ìm filter, pressure regulator (Depending on Model), and power cord.

Key Features

  • Display can be tilted for optimal reading
  • Variable-flow control for dispensing of Type I water
  • Quick-change cartridge design simplifies self maintenance, even in mid-operations
  • 1 year warranty

More About this Item

Barnstead Smart2Pure systems deliver up to 72 L/day of Type I ultrapure water or Type II pure water from a tap feed water supply. Depending on your requirements, choose from three production capacities—3, 6, or 12 L/hr. The 12 L/hr systems let you safely store high purity water in either a 30- or 60-L reservoir, easily accessible when you need it!

These compact systems are fully customizable to support your tissue culture, PCR, electrophoresis, atomic absorption, ICP, ICP/MS, and chromatography needs. In addition, applications requiring Type II water such as reagent preparation and glassware rinsing can be met. The RO membrane with pretreatment cartridge is separate from the DI cartridge, ensuring you only replace each cartridge as needed.

The systems with UV option offer UV-oxidation (185/254 nm) for reducing the content of micoorganisms and their metabolites. The systems with UF option include an ultrafilter to eliminate endotoxins.

An internal 6-L tank with vent filter is included.

Pretreatment filter is required (sold seperately)

Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.

1 Year WarrantyConformite EuropeenneCanadian Standards Association
Specifications
  • ASTM Water GradeType I and Type II
  • Max Production (L/hour)6
  • Max Flow Rate (L/min)1
  • Inlet Port1/4" OD compression
  • With UV LampYes
  • With UltrafilterYes
  • Feed Water Requirements (Source)Potable Water according to DIN 2000
  • Type I Water (Resistivity)18.2 MΩ-cm
  • Type I Water (TOC)1 to 5 ppb
  • Type I Water (Conductivity)0.055 µS/cm
  • Feed Water Requirements (pH range)4 to 11
  • Feed Water Requirements (pressure)1.4 to 87 psi (0.1 to 6 bar)
  • Type II Water (Resistivity)15 to 10 MΩ-cm
  • Feed Water Requirements (Temperature)2-35⁰C
  • Type II Water (Conductivity)0.067 to 0.1 µS-cm
  • Min Operating Pressure (PSI)29
  • Max Operating Pressure (PSI)87
  • Min Operating Pressure (bar)2
  • Max Operating Pressure (bar)6
  • Height (in)15.75
  • Depth (in)12
  • Width (in)21.5
  • Height (cm)30.5
  • Depth (cm)54.5
  • Width (cm)40
  • DescriptionSmart2Pure 6 l/h UV/UF
  • Warranty1 year
Reviews
Q&A