Testo 115i Smart Probe Clamp Thermometer for use with Smartphone or Tablet
Representative image only.
Testo 115i Smart Probe Clamp Thermometer for use with Smartphone or Tablet
Testo | Mfr: 0560 2115 03| Item: 3000918
View HVAC measurement on your Smart device from up to 60 feet away!
$190.07CAD / Each
$190.07CAD / Each
Buy It Together
Current Item
Testo 115i Smart Probe Clamp Thermometer for use with Smartphone or Tablet
Testo 115i Smart Probe Clamp Thermometer for use with Smartphone or Tablet
3000918
$190.07CAD / Each
+
Testo Smart Probe AC/R Case
3000949
$88.40CAD / Each
=
Total Price 278.47 ADD 2 ITEMS TO CART
Product Description
Includes
Three AAA batteries and certificate of calibration.
Key Features
  • Use the Smart Probes app to read and create custom reports
  • Save and share measurements as a PDF or Excel® file
  • Perform difficult calculations automatically
  • Multi-color LED displays low battery warning and Bluetooth® connectivity
  • Grips pipes up to 1.5" in diameter
  • 2 year warranty
More About this Item
This compact, rugged, easy-to-use wireless clamp thermometer, designed to grip pipes up to 1.5” in diameter, provides accurate temperature measurements of pipes in heating and cooling systems – ideal for the HVAC professional.
Easily monitor and transmit all measured data wirelessly via Bluetooth® to your smartphone or tablet from up to 60 feet away. With the free Testo Smart Probes app, you can display measurements from up to six Smart Probes at the same time, visualize measurement value changes as a graph or a table, and perform automatic calculations. Easily turn your data into a custom report, attach images created with the app, and send via email as a PDF or Excel® file.
Monitor superheating and subcooling in AC and refrigeration systems by using two 115i clamp thermometer probes in conjunction with two 549i Smart Probe high-pressure refrigeration probes. The clamp thermometer can also be used in conjunction with the 510i Smart Probe differential pressure manometer and 805i Smart Probe infrared thermometer to monitor hydronic heating systems.
Available for sale in the United States and Canada only.
Two year warrantyApple App StoreConformite EuropeenneGoogle Play App
Specifications
  • Min Temperature (° F)-40
  • Max Temperature (° F)302
  • Min Temperature (° C)-40
  • Max Temperature (° C)150
  • Accuracy±2.3°F (1.3°C) from –4 to 185°F (–20 to 85°C)
  • Resolution0.1°
  • Probe TypeNTC clamp temperature probe
  • Low Battery IndicatorYes
  • Battery3 AAA (included)
  • DescriptionSmart Probe Clamp Thermometer for use with Smartphone or Tablet, NTC