Terracon Rinse Station, Floor Model

Representative image only.

Terracon Rinse Station, Floor Model

Terracon | Mfr: CTS1444| Item: 9896508

Compact design keeps water contained – no more wet floors and slippery hazards!

$19,411.91CAD / Each

$19,411.91CAD / Each

Buy It Together

Current Item

Terracon Rinse Station, Floor Model
Terracon Rinse Station, Floor Model
9896508

$19,411.91CAD / Each
+ =
Total Price 43111.39 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Choose from countertop or floor models
  • Avoid expensive automated systems
  • Eliminate manual cleaning efforts
  • 1 year warranty

More About this Item

These cost-effective rinse stations clean all sizes and styles of vessels and carboys up to 50 gallons (200 liters). The rinse station is easy to use and sets up quickly for use. No electricity is needed — station connects directly to water inlet and drain. The top panel of the rinse station is manufactured with roto-molded polyethelyne, and base and legs are manufactured with fabricated polyethylene.


Countertop unit will take vessels in diameter up to 15" (38.1 cm). Rinse is turned on and off by a hand-controlled valve on the side of the unit. The floor model unit will take vessels with diameter up to 19" (18.3 cm). Rinse is turned on and off by a foot-controlled valve attached to the bottom platform of the unit. Both valves are made of polypropylene with Viton™ elastomers and a Kynar™ coated stainless steel spring. Spray head is manufactured with PVDF. The inlet water connection is 1/2" NPT(F); drain connection is 1 1/2" NPT(F).


Special connections and customized units are available — contact an Application Specialist for more information.

1 Year Warranty
Specifications
  • Exterior Width (in)22
  • Exterior Height (in)53
  • Exterior Depth (in)22
  • Inlet Connection1/2" NPT(F)
  • Outlet Connections1-1/2" NPT(F)
  • Shipping Weight (lbs)36
  • DescriptionRinse Station, Floor Model
  • Warranty1 Year
Reviews
Q&A