Tear Away Lid for 2 Gal. Pail 06274-03
Representative image only.
Tear Away Lid for 2 Gal. Pail 06274-03
| Item: 0627434
A "tear-tab" strip allows easy access to product
$7.22CAD / Each
BUY MORE & SAVE
  • 1-5$7.22 CAD / Each
  • 6-29$6.50 CAD / Each
  • 30-119$6.14 CAD / Each
  • 120+$5.78 CAD / Each
$7.22CAD / Each
BUY MORE & SAVE
  • 1-5$7.22 CAD / Each
  • 6-29$6.50 CAD / Each
  • 30-119$6.14 CAD / Each
  • 120+$5.78 CAD / Each
Buy It Together
Current Item
Tear Away Lid for 2 Gal. Pail 06274-03
Tear Away Lid for 2 Gal. Pail 06274-03
0627434
$7.22CAD / Each
+
Heavy-Duty Smart Container, HDPE, 30 gal
0674050
$282.89CAD / Each
=
Total Price 290.11 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Made from FDA approved HDPE
  • No lid opener required
  • Nests inside another for easy storage
  • Meets FDA 21 CFR 177.1520(c) 3.1a
  • Compatible with 2 gallon pail 06274-03
More About this Item
Environmentally friendly, fully recyclable covers are made from FDA approved HDPE. Easily open lid without using tools.
Specifications
  • MaterialHDPE
  • ColorWhite
  • Compatible WithCompatible with 2 gallon pail 06274-03
  • Diameter (InchShort)9 15/16
  • DescriptionTear Away Lid for 2 Gal. Pail 06274-03