Taylor 9878 Pen Style Digital Pocket Thermometer with Large Head
Representative image only.
Taylor 9878 Pen Style Digital Pocket Thermometer with Large Head
Taylor | Mfr: 9878E| Item: 9000020
Ideal for use in the food industry and all other general applications
$55.99CAD / Each
BUY MORE & SAVE
  • 1-11$55.99 CAD / Each
  • 12+$50.39 CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$55.99CAD / Each
BUY MORE & SAVE
  • 1-11$55.99 CAD / Each
  • 12+$50.39 CAD / Each
Buy It Together
Current Item
Taylor 9878 Pen Style Digital Pocket Thermometer with Large Head
Taylor 9878 Pen Style Digital Pocket Thermometer with Large Head
9000020
$55.99CAD / Each
+ =
Total Price 208.85 ADD 2 ITEMS TO CART
Product Description
Includes
storage sleeve with clip and 1 CR2032 battery.
Key Features
  • Maintain product safety with accurate readings
  • -40 to 500 degree F temperature range
  • 1.5mm dia. stepdown probe meets FDA recommendation
  • Maximum and minimum temperature recall features
  • Temperature Hold feature
  • Can be recalibrated
  • Antimicrobial plastic storage case slows the growth of mold and bacteria
  • 4.5-in. stainless steel stem
  • Waterproof design
  • Auto off feature helps conserve battery and can be disabled
  • 1 year warranty
More About this Item
The real secret to great cooking is a Taylor thermometer. Thermometers are the only way to ensure that foods are cooked to the proper temperature, your oven is calibrated to the correct degree, and that your refrigerator is keeping your food cold and safe. Taylor is and has been the leader in food thermometers, with expertise over 150 years strong. You can count on Taylor for accuracy, superior performance, and dependability. This is why Taylor has tools chefs trust
This Taylor 9878E digital probe thermometer is perfect for keeping in your pocket when working in your busy commercial kitchen. The simple, compact design isn't the only thing to love, either. This thermometer is packed with features, including a backlight display; auto-off, min/max registry, and hold buttons; as well as an anti-microbial housing and sleeve that prevents bacterial growth.
The FDA compliant probe on this Taylor thermometer measures 4 1/2" in length and 1.5mm in diameter, making it ideal for use with thin foods like burgers and fish fillets. It's able to be re-calibrated for maximum accuracy and compliance with HACCP plans.
When it comes to sanitation, this thermometer is the clear choice. It's completely waterproof and safe for hand washing with the rest of your kitchen tools. A CR2032 replaceable lithium battery is included
1 Year Warranty
Specifications
  • Meter Measurement TypeDigital Indicator
  • Number Of Probe Channels1
  • Min Temperature (° F)-58
  • Max Temperature (° F)500
  • Min Temperature (° C)-50
  • Max Temperature (° C)260
  • Accuracy±2.0°F (±1.0°C)
  • Resolution0.1°
  • Display Type4-digit LCD
  • Height (in)19/64
  • No. of Probe(s) Included1
  • Probe Diameter (in)0.14
  • Probe Length (in)4 51/64
  • Battery1 1.5V Lithium (included)
  • DescriptionPen Style Digital Pocket Thermometer with Large Head