Taylor 5983N Class Series Candy / Deep Fry Analog Glass Tube Thermometer with Eteched Temperature Zones in Glass
Representative image only.
Taylor 5983N Class Series Candy / Deep Fry Analog Glass Tube Thermometer with Eteched Temperature Zones in Glass
Taylor | Mfr: 5983N| Item: 9000137
Use this thermometer to ensure a consistent final product every time!
$29.03CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$29.03CAD / Each
Buy It Together
Current Item
Taylor 5983N Class Series Candy / Deep Fry Analog Glass Tube Thermometer with Eteched Temperature Zones in Glass
Taylor 5983N Class Series Candy / Deep Fry Analog Glass Tube Thermometer with Eteched Temperature Zones in Glass
9000137
$29.03CAD / Each
+ =
Total Price 181.89 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Adjust immersion depth with the included clip
  • Large, easy-to-read to read temperature zones.
  • Temperture range 100 to 400°F (40°C to 200°C)
  • Spirit filled glass thermometer tube.
  • Common candy temperatures etched into casing.
  • Insulated handle with built-in pan clip.
  • Durable stainless steel construction..
  • 1 year warranty
More About this Item
An essential addition to commercial kitchens, this thermometer allows you to easily check the temperatures of delicate candy mixes or fryer oils so your final food products come out just right.
he 5983N Taylor Candy/Deep Fry Thermometer is designed to be easy to hold and easy to read, and it can easily rest in a pot nicely to give an accurate temperature reading when you need it.
This candy thermometer is 12 inches long, so it can sink deep enough into a pot for an accurate reading. The thermometer’s length also leaves plenty of room for large numbers, that are easy to read while you are cooking over a hot stove. The large, visible temperature zones are marked plainly beginning at 100 degrees Fahrenheit and going as high as 400 degrees. On the right you will find Celsius temperature markings as well as critical stages for candy making including indicators for soft ball, hard ball, soft crack and hard crack stages. The glass thermometer tube operates with alcohol instead of mercury, making this a good choice for cooks who are concerned about using the toxic substance around food. With common candy temperatures etched into the thermometer’s metal casing, it is easy to tell when your concoction has reached the appropriate heat stage, and you won’t have to remove or jostle the thermometer to get your reading. This thermometer also boasts an oversized, insulated handle. Its size and shape make the handle easy to grab and hold even when you are wearing a heat glove. At the back of the handle, you will find a built-in pan clip to help keep the thermometer safe and steady while you are cooking. The stainless steel construction of the 5983N Taylor Candy/Deep Fry Thermometer makes it a durable kitchen instrument and easy to clean. The shape of the thermometer precludes using it as a probe, however; so you won’t be able to insert the tip into anything but liquid. Overall, this thermometer is a handy and easy-to-use kitchen device that candy makers will appreciate for its clear, large temperature gauge and clearly marked candy making temperature ranges. The overall length makes the 5983N Taylor Candy/Deep Fry Thermometer a smart choice for people who make large batches of candy in deep pot
1 Year Warranty
Specifications
  • Min Temperature (° F)100
  • Max Temperature (° F)400
  • Min Temperature (° C)40
  • Max Temperature (° C)200
  • Accuracy2%
  • Stem Length (in)12
  • DescriptionCandy / Deep Fry Analog Glass Tube Thermometer with Eteched Temperature Zones in Glass