Taylor 5932 Oven / Grill Analog Dial Thermometer with Large Face and Dual Scale
Representative image only.
Taylor 5932 Oven / Grill Analog Dial Thermometer with Large Face and Dual Scale
Taylor | Mfr: 5932| Item: 9000052
Get more reliable, consistent results
$13.11CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$13.11CAD / Each
Buy It Together
Current Item
Taylor 5932 Oven / Grill Analog Dial Thermometer with Large Face and Dual Scale
Taylor 5932 Oven / Grill Analog Dial Thermometer with Large Face and Dual Scale
9000052
$13.11CAD / Each
+ =
Total Price 165.97 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Extra large, colorful 3 1/4" dial.
  • Temperature range 100 to 600°F (50 to 300°C)
  • Red pointer.
  • Glass lens.
  • Durable stainless steel casing.
  • Hangs or stands.
  • NSF listed.
  • 5 year warranty
More About this Item
Bimetal Oven Thermometer features an extra-large 3-1/4" (8.3 cm) dual-scale dial with a durable stainless steel casing and glass lens. Built-in hanger and stand works in any oven. Leave in oven to monitor temperatures. Hand wash only.
5 Year Warranty
Specifications
  • Min Temperature (° F)100
  • Min Temperature (° C)38
  • Max Temperature (° F)600
  • Max Temperature (° C)316
  • Accuracy±10°F
  • Division (° C)5
  • Division (° F)5
  • Dial Size (in)2.75
  • Stem Material304 Stainless Steel
  • DescriptionOven / Grill Analog Dial Thermometer with Large Face and Dual Scale
  • Warranty5 years