Swift Optical M3-B Micro/Macro Cordless Field Microscope, Binocular
Representative image only.
Swift Optical M3-B Micro/Macro Cordless Field Microscope, Binocular
Swift Optical | Mfr: M3-B| Item: 0310026
Cordless microscope uses rechargeable batteries for easy field use
$1,084.42CAD / Each
$1,084.42CAD / Each
Product Description
Includes
specimen cup, contrast plate, stage plate, rechargeable batteries, and AC adapter.
Key Features
  • Versatile design allows for use as a microscope, macroscope, or in the field
  • Sturdy detachable tripod legs fold for easy storage and sturdy even on rugged outdoor surfaces
  • Energy efficient, rechargeable LED illumination allows up to 40 hours of use on a single 8 hour charge
  • 3 year warranty
More About this Item
Rechargeable batteries, tripod legs, and a carrying handle make this microscope ideal for field use — take it wherever you need it! Durable all-metal construction withstands outdoor environments. Versatile microscope features three objective lenses for microscopic use and one lens for macroscopic viewing of larger objects. Top and bottom LED illumination uses little power allowing up to 40 hours of field use before recharging is necessary. Stage position adjusts for micro- and macro-viewing. Tripod legs fold for easy storage; for added versatility, microscope can be mounted on a standard camera tripod for easier field use.
3 Year Warranty
Specifications
  • Magnification20x/40x/100x/400x
  • Maximum Magnification400x
  • Head ConfigurationBinocular
  • Working Distance18 mm
  • Illuminator(s)LED, top and bottom
  • PowerRechargeable batteries and VAC adapter
  • Power (VAC)110
  • DescriptionMicro/Macro Cordless Field Microscope, Binocular
  • Warranty3 Years