Sussman 39183F Replacment Heating Element, 18 kW, 480 VAC

Representative image only.

Sussman 39183F Replacment Heating Element, 18 kW, 480 VAC

Sussman | Mfr: 39183F| Item: 0760353

$1,371.73CAD / Each

$1,371.73CAD / Each
Product Description

Key Features

  • 1 year warranty

More About this Item

Keep your steam generator running smoothly with these parts and accessories.

1 Year Warranty
Specifications
  • Compatible WithSussman Steam Generator with 18 kW, 480 VAC power
  • Power (VAC)480
  • DescriptionReplacment Heating Element, 18 kW, 480 VAC
  • Warranty1 year
Reviews
Q&A