Sussman 39183C Replacment Heating Element, 18 kW, 240 VAC

Representative image only.

Sussman 39183C Replacment Heating Element, 18 kW, 240 VAC

Sussman | Mfr: 39183C| Item: 0760351

$1,371.73CAD / Each

$1,371.73CAD / Each
Product Description

Key Features

  • 1 year warranty

More About this Item

Keep your steam generator running smoothly with these parts and accessories.

1 Year Warranty
Specifications
  • Compatible WithSussman Steam Generator with 18 kW, 240 VAC power
  • Power (VAC)240
  • DescriptionReplacment Heating Element, 18 kW, 240 VAC
  • Warranty1 year
Reviews
Q&A