Stony Brook PDVDI-120 Portable Falling Needle Field Viscometer 98939-30

Representative image only.

Stony Brook PDVDI-120 Portable Falling Needle Field Viscometer 98939-30

Stony Brook | Mfr: PDVDI-120| Item: 9893930

Ease of use and compact size makes this viscometer perfect for portable/field applications

$1,694.03CAD / Each

$1,694.03CAD / Each
Product Description

Includes

viscometer main body, two reusable sample tubes, falling needle, stopwatch, digital thermometer, and carrying case.

Key Features

  • Conforms to SAE AIR and ASTM D5478
  • Includes everything you need to start taking measurements
  • 1 year warranty

More About this Item

Take viscosity measurements in the field or on the plant floor for Newtonian and non-Newtonian applications including adhesives, dairy products, deicing fluids, honey, ketchup, paints, petroleum products, surfactants, and more! Simply pour your sample into the tube, insert and drop the needle, and measure the falling time between two markings. The falling time is measured manually with the included stopwatch. Take multiple measurements with the same sample and take an average to decrease human error.

1 Year Warranty
Specifications
  • Min Viscosity Range (cP)2
  • Max Viscosity Range (cP)1000000
  • Viscosity Repeatability±2%
  • Viscosity Accuracy±2%
  • Sample Volume Needed (mL)25
  • DescriptionPortable Falling Needle Field Viscometer 98939-30
  • Warranty1 year
Reviews
Q&A