Starna Scientific DMV-BioCell Cuvette, 2.5 µl, Z Height 15 mm

Representative image only.

Starna Scientific DMV-BioCell Cuvette, 2.5 µl, Z Height 15 mm

| Mfr: 83056-75| Item: 8305675

Direct low volume analysis of nucleic acids and proteins

$4,840.09CAD / Each

$4,840.09CAD / Each
Product Description

Includes

DNACON verification standard.

Key Features

  • High accuracy and reproducibility for measurements of nucleic acids and proteins
  • Uses conventional sample position of a spectrophotometer – no special holder or positioning required
  • Optical path can be validated to a high degree of accuracy with DNACON certified reference materials
  • Magnetic closure design facilitates rapid filling and emptying and easy cleaning

More About this Item

Compatible with all major UV/Visible spectrophotometers the DMV-Bio Cell offers high accuracy and reproducibility for direct, low volume measurements of nucleic acids or proteins without the need for a dedicated system. This cuvette features no integral reflective surfaces to keep clean and a window for sample viewing. Included with certified reference materials for validated measurements to give truly validated measurements and the ultimate confidence in results.

When combined with the Jenway Genova Bio spectrophotometer (83056-76), this system offers an elegant solution to direct, accurate measurement of nucleic acids or proteins in low volume biological samples. Spectrophotometer sold separately.

Specifications
  • Max Capacity (MicroL)2.5
  • Top TypeMagnetic closure
  • Pathlength (mm)0.5
  • Beam Height Zd (mm)15
  • Height (in)2 2/5
  • Width (in)1/2
  • Depth (in)1/2
  • Height (mm)61
  • Width (mm)12.5
  • Depth (mm)12.5
  • DescriptionDMV-BioCell Cuvette, 2.5 µl, Z Height 15 mm
Documents & Certificates
Reviews
Q&A