Spectra Por 131057 Biotech-Grade Dialysis Tubing, 0.1-0.5 kDalton, 24mm, 33ft/roll

Representative image only.

Spectra Por 131057 Biotech-Grade Dialysis Tubing, 0.1-0.5 kDalton, 24mm, 33ft/roll

Spectra Por | Mfr: 131057| Item: 0289088

Ideal for concentration, purification, and desalination

$1,100.62CAD / Box of 1

$1,100.62CAD / Box of 1

Buy It Together

Current Item

Spectra Por 131057 Biotech-Grade Dialysis Tubing, 0.1-0.5 kDalton, 24mm, 33ft/roll
Spectra Por 131057 Biotech-Grade Dialysis Tubing, 0.1-0.5 kDalton, 24mm, 33ft/roll
0289088

$1,100.62CAD / Box of 1
+ + + =
Total Price 2062.38 ADD 4 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • High-purity membranes with no heavy metal or sulfide contaminants
  • Low protein adsorption
  • 1 year warranty

More About this Item

Cellulose Ester (CE) offers a wider range of more precise MWCO’s for critical separations; packaged as a 10-m roll in 0.05% sodium azide.

1 Year Warranty
Specifications
  • MaterialCellulose Ester, biotech-grade
  • Max Molecular Weight Cut Off (Mwco) (Dalton)500
  • Min Molecular Weight Cut Off (Mwco) (Dalton)100
  • Flat Width (mm)24
  • Diameter (mm)15
  • Length Per Pack (ft)33
  • Diameter (in)0.591
  • DescriptionBiotech-Grade Dialysis Tubing, 0.1-0.5 kDalton, 24mm, 33ft/roll
  • Warranty1 year
Reviews
Q&A