SP Wilmad High-Throughput NMR Tube and Cap, 4.94665 mm OD x 7" L, Large Marking Spot; 100/Pk

Representative image only.

SP Wilmad High-Throughput NMR Tube and Cap, 4.94665 mm OD x 7" L, Large Marking Spot; 100/Pk

SP Wilmad | Mfr: WG-1003-7| Item: 8399961

High-throughput NMR tubes have one of the best OD tolerances in the industry

$265.80CAD / Pkg of 100

$265.80CAD / Pkg of 100
Product Description

Includes

NMR tubes with marking spot and caps.

Key Features

  • NMR tubes are constructed of ASTM E438 Type 1 Class B borosilicate glass
  • Ensure spectrum clarity with an averaged camber of 60 micron
  • NMR tube features a 7" length, 4.94665 mm OD, and 0.43 mm wall thickness

More About this Item

These Wilmad-LabGlass high-throughput NMR tubes have an averaged camber of 60 micron to guarantee the spectrum resolution for small molecules (MW<250) samples up to 600 MHz. Each 5mm tube meets precise wall thickness to ensure they fit tightly in spinner turbine. All tubes are made from borosilicate glass. Choose from tubes with authentic Bruker® SampleJet® caps without code or tubes with disposable caps.

Specifications
  • SterileNo
  • Tube MaterialGlass
  • Tube Length (mm)178
  • Tube Outside Diameter (OD) (mm)4.94665
  • Cap IncludedYes
  • Cap StyleDisposable
  • GraduationsLarge Marking Spot
  • Special Features600 MHz
  • DescriptionHigh-Throughput NMR Tube and Cap, 4.94665 mm OD x 7" L, Large Marking Spot; 100/Pk
Reviews
Q&A