SP Wilmad Bruker® SampleJet® High-Throughput NMR Tube and Cap, 5 mm OD x 103.5 mm L; 100/Pk

Representative image only.

SP Wilmad Bruker® SampleJet® High-Throughput NMR Tube and Cap, 5 mm OD x 103.5 mm L; 100/Pk

SP Wilmad | Mfr: WG-1000-4-SJ| Item: 8399963

High-throughput NMR tubes have one of the best OD tolerances in the industry

$329.45CAD / Pkg of 100

$329.45CAD / Pkg of 100
Product Description

Includes

Bruker® SampleJet® NMR tubes and Bruker® SampleJet® caps.

Key Features

  • NMR tubes are constructed of ASTM E438 Type 1 Class B borosilicate glass
  • Ensure spectrum clarity with an averaged camber of 60 micron
  • NMR tube features a 103.5 mm length, 5 mm OD, and 0.43 mm wall thickness

More About this Item

These Wilmad-LabGlass high-throughput NMR tubes have an averaged camber of 60 micron to guarantee the spectrum resolution for small molecules (MW<250) samples up to 600 MHz. Each 5mm tube meets precise wall thickness to ensure they fit tightly in spinner turbine. All tubes are made from borosilicate glass. Choose from tubes with authentic Bruker® SampleJet® caps without code or tubes with disposable caps.

Specifications
  • SterileNo
  • Tube MaterialGlass
  • Tube Length (mm)103.5
  • Tube Outside Diameter (OD) (mm)5
  • Cap IncludedYes
  • Cap StyleBruker® SampleJet®
  • GraduationsNo
  • Special Features600 MHz
  • DescriptionBruker® SampleJet® High-Throughput NMR Tube and Cap, 5 mm OD x 103.5 mm L; 100/Pk
Reviews
Q&A