SP Hotpack Glassware Washer, Freestanding, with Tap Water Rinse, Top and Bottom Standard Racks; 115V 60 Hz

Representative image only.

SP Hotpack Glassware Washer, Freestanding, with Tap Water Rinse, Top and Bottom Standard Racks; 115V 60 Hz

| Mfr: UCW-HA00101| Item: 7890354

Efficient, minimal water consumption with spotless results

$13,813.03CAD / Each

$13,813.03CAD / Each

Buy It Together

Current Item

SP Hotpack Glassware Washer, Freestanding, with Tap Water Rinse, Top and Bottom Standard Racks; 115V 60 Hz
SP Hotpack Glassware Washer, Freestanding, with Tap Water Rinse, Top and Bottom Standard Racks; 115V 60 Hz
7890354

$13,813.03CAD / Each
+ +
SP Hotpack DI Water Boost Pump for Glassware Washer
7890362

$1,532.33CAD / Each
+ =
Total Price 21585.16 ADD 4 ITEMS TO CART

Customers Also Bought

Product Description

Includes

top and bottom stainless steel open racks.

Key Features

  • Designed and insulated for quiet operation
  • Optimize temperature and time for specific cleaning with 6-cycle selection
  • Enhanced dual fan drying system draws in dry air and exhausts moist air out
  • Ensure long life and corrosion protection with 304 stainless steel chamber
  • Delayed start feature allows washer to operate unattended during off-peak hours to reduce energy costs
  • 1 year warranty

More About this Item

Designed for ease of operation and flexible loading, these SP Hotpack® undercounter and freestanding glassware washers feature upper and lower open racks and can accommodate a wide range of glassware. All models feature a soft touch controller, improved dryer, and noise buffering insulation for a quieter run. Three separate filters protect pumps from debris and are conveniently located for easy access. Multiple cycle selections and microprocessor control systems ensure that each load is processed with minimum energy consumption. Six cycle selections let you select optimal temperature and include a customizable timed option. These 6 cycles include rinse only, normal wash, daily wash, heavy wash, heavy wash time saver, and timed programs.

Choose from undercounter and freestanding models with standard, DI rinse, and DI rinse/spindle rack ready features. Undercounter units have adjustable height leveling feet and freestanding units have locking casters for portability. All DI models use facility-provided deionized water for final rinse and have the option of a DI booster pump (sold separately), recommended when the incoming DI water source has a pressure of less than 18 psi. Spindle-ready models can accommodate direct injection spindle racks (sold separately) for narrow-neck glassware.

Canadian Standards Association1 Year Warranty
Specifications
  • Water Consumption (Gal)4
  • Exterior Width (in)24
  • Exterior Height (in)38.5
  • Exterior Depth (in)24.25
  • Power (VAC)115
  • Power (Hz)60
  • Washing Temperature (° F)158
  • Washing Temperature (° C)70
  • Shipping Weight (lbs)136.5
  • DescriptionGlassware Washer, Freestanding, with Tap Water Rinse and Top and Bottom Standard Racks; 115V 60 Hz
  • Warranty1 Year
Reviews
Q&A