Smith & Wesson SW152PCCMINII Safety Glasses, Clear Lens
Representative image only.
Smith & Wesson SW152PCCMINII Safety Glasses, Clear Lens
Smith & Wesson | Mfr: 19822| Item: 8628600
Adjustable temples have four length settings—easily customize them for a secure fit
$20.63CAD / Each
BUY MORE & SAVE
  • 1-9$20.63 CAD / Each
  • 10+$18.57 CAD / Each
$20.63CAD / Each
BUY MORE & SAVE
  • 1-9$20.63 CAD / Each
  • 10+$18.57 CAD / Each
Product Description
Key Features
  • Ribbed, nonslip rubber nose-piece provides for all-day comfort
More About this Item
Mini-Magnum™ glasses are styled to fit individuals who require a smaller version.
Compliance: ANSI Z87.1+ (High impact)
Specifications
  • Frame ColorBlack
  • Lens ColorClear
  • Lens CoatingNone
  • ComplianceANSI Z87.1+ (High Impact)
  • DescriptionSafety Glasses, Clear Lens