Smith & Wesson 3016306 Safety Glasses, Gun Metal frame, Clear lens
Representative image only.
Smith & Wesson 3016306 Safety Glasses, Gun Metal frame, Clear lens
Smith & Wesson | Mfr: 21294| Item: 8649401
Stylized frames feature a cutting-edge design that's ideal for work and play
$20.63CAD / Each
BUY MORE & SAVE
  • 1-11$20.63 CAD / Each
  • 12+$18.57 CAD / Each
$20.63CAD / Each
BUY MORE & SAVE
  • 1-11$20.63 CAD / Each
  • 12+$18.57 CAD / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Glasses feature soft rubber nose pads for superior comfort and fit. Sport-grip temples eliminate slippage. Dual lenses with an 8-base curve provide wraparound protection. High-performance polycarbonate lens provides 99.9% UV protection. Includes a micro-fiber carry pouch that doubles as a cleaning cloth.
Compliance: ANSI Z87.1+-2003
1 Year Warranty
Specifications
  • Frame ColorGunmetal
  • Lens ColorClear
  • ComplianceANSI Z87.1+ (High Impact)
  • DescriptionSafety Glasses, Gun Metal frame, Clear lens