SHOWA 910-08 Cut-Resistant Glove, Uncoated; size, medium
Representative image only.
SHOWA 910-08 Cut-Resistant Glove, Uncoated; size, medium
SHOWA | Mfr: 910-08| Item: 8628951
Safely handle glass, sheet and scrap metal, or process food
$16.21CAD / Each
BUY MORE & SAVE
  • 1-11$16.21 CAD / Each
  • 12+$14.59 CAD / Each
$16.21CAD / Each
BUY MORE & SAVE
  • 1-11$16.21 CAD / Each
  • 12+$14.59 CAD / Each
Product Description
More About this Item
Lightweight 10-gauge seamless knit glove offers dexterity, tactile sensitivity, and protection. Use to safely handle glass, sheet metal, scrap metal, and to process food. Glove offer superior cut protection with a special wrapping process using Dynema® fibers. Washable and bleachable. Not designed for use with moving or serrated blades. Ambidextrous.
Approval: CE
Compliance: 21CFR, 170-199, acceptable for food and pharmaceutical use.
Conformite Europeenne
Specifications
  • MaterialDyneema®
  • Thickness (mil)10
  • SizeMedium
  • ColorWhite
  • ComplianceFDA 21 CFR 170-199
  • SterileNo
  • DescriptionCut-Resistant Glove, Uncoated; size, medium