SHOWA 890-10 Viton Gloves, extra-large, 28-mil thickness, 1 pair
Representative image only.
SHOWA 890-10 Viton Gloves, extra-large, 28-mil thickness, 1 pair
SHOWA | Mfr: 890-10| Item: 8628833
Get flexible, heavy-duty protection
$521.58CAD / Pair of 1
BUY MORE & SAVE
  • 1-5$521.58 CAD / Pair of 1
  • 6+$469.42 CAD / Pair of 1
$521.58CAD / Pair of 1
BUY MORE & SAVE
  • 1-5$521.58 CAD / Pair of 1
  • 6+$469.42 CAD / Pair of 1
Buy It Together
Current Item
SHOWA 890-10 Viton Gloves, extra-large, 28-mil thickness, 1 pair
SHOWA 890-10 Viton Gloves, extra-large, 28-mil thickness, 1 pair
8628833
$521.58CAD / Pair of 1
+
SHOWA 874-08 Gloves, smooth, medium
8635701
$148.44CAD / Pair of 1
=
Total Price 670.02 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Superior resistance to PCBs
More About this Item
Made of Viton® fluoroelastomer, these gloves provide superior resistance to PCBs, chlorinated and aromatic solvents, and vapors. Smooth, flexible, unlined gloves with a rolled cuff are injection-molded for consistent thickness throughout.
Specifications
  • MaterialViton
  • Thickness (mil)30
  • SizeX-Large
  • Length (in)14
  • Length (cm)35
  • ColorBlack
  • SterileNo
  • DescriptionViton Gloves, extra-large, 28-mil thickness, 1 pair