Shimpo ST-320BL-2 Black Light LED Stroboscope; Rechargeable Battery
Representative image only.
Shimpo ST-320BL-2 Black Light LED Stroboscope; Rechargeable Battery
Shimpo | Mfr: ST-320BL-2| Item: 5350219
Illuminate UV inks used in labels, watermarks, security tags, and more
$1,908.04CAD / Each
for an additional
$307.57CAD / Each
for an additional
$432.44CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$1,908.04CAD / Each
Buy It Together
Current Item
Shimpo ST-320BL-2 Black Light LED Stroboscope; Rechargeable Battery
Shimpo ST-320BL-2 Black Light LED Stroboscope; Rechargeable Battery
5350219
$1,908.04CAD / Each
+
InnoCal NIST-Traceable Calibration; Stroboscope
1705010
$307.57CAD / Each
=
Total Price 2215.61 ADD 2 ITEMS TO CART
Product Description
Includes
One rechargeable Lithium battery and power adapter.
Key Features
  • Versatile design — use portably or mount permanently
  • Use in almost any high-speed application with the wide flash range
  • Obtain reliable use from the long-life LED black light lamps — three to five year life-expectancy
  • Remove trigger handle to expose threads for mounted use
  • Allow process machinery to control flash rate with the input trigger function
  • 1 year warranty
More About this Item
The ST-320BL stroboscope features nine high-powered, black light CREE LEDs — ideal for use in UV printing, packaging, textile, automotive, cable, mining, steel, chemical, optical, and medical industries for speed and frequency measurements. The wide flash range covers 60 to 120,000 flashes per minute (1 to 2000 Hz) and may be adjusted for sharper images and detailed inspections. The input trigger function allows process machinery to control the flash rate for automatic adjustments to process fluctuations.
What's included: a rechargeable battery, universal (100 to 240 VAC) charger, and foam-lined metal carrying case.
1 Year WarrantyConformite EuropeenneIP54 Rated
Specifications
  • Min Flash Range (fpm)60
  • Max Flash Range (fpm)120000
  • Flash Resolution (fpm)0.1 from 60 to 12,000 fpm; 1 from 12,001 to 120,000 fpm
  • Flash Accuracy (fpm)0.01% ±1 digit full-scale at 77°F (25°C)
  • Input12 V pulse input (3 pin M-12)
  • Display Type6-digit backlit LCD
  • Battery Type1 Rechargeable Lithium (included)
  • DescriptionBlack Light LED Stroboscope; Rechargeable Battery