Sensidyne 830-0001-01 Gas Sampling Pump, 50 or 100 mL sample volumes
Representative image only.
Sensidyne 830-0001-01 Gas Sampling Pump, 50 or 100 mL sample volumes
Sensidyne | Mfr: 830-0001-01| Item: 8190005
Ensure accurate sample taking with a single stroke
$826.57CAD / Each
$826.57CAD / Each
Product Description
Key Features
  • Fail-safe handle lock ensures accurate 50- and 100-cc samples with a single stroke
  • Limited Lifetime Warranty
More About this Item
Precision, hand-operated piston pump works with Sensidyne® gas detection tubes (sold separately). Features stroke counter, flow-finish indicator that signals stroke completion, and built-in tube-tip breaker. Includes soft case.
Limited Lifetime Warranty
Specifications
  • DescriptionGas Sampling Pump, 50 or 100 mL sample volumes
  • WarrantyLimited Lifetime